Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HuC Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB092383 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092383 100 μL
NB092384 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092383 Supplier Novus Biologicals Supplier No. NBP302943100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HuC Polyclonal specifically detects HuC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HuC
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DKFZp547J036, ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C), ELAV-like protein 3, Hu antigen C, Hu-antigen C, HuC, HUCL, HUCParaneoplastic cerebellar degeneration-associated antigen, MGC20653, PLE21Paraneoplastic limbic encephalitis antigen 21
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of humanHuC (NP_001411.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1995
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.