Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human 14-3-3 epsilon Recombinant Protein
SDP

Catalog No. p-7112726 Shop All Novus Biologicals Products
Click to view available options
Quantity:
100 μg
20 μg

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-255 of Human 14-3-3 epsilon The Recombinant Human 14-3-3 epsilon Protein is derived from E. coli. The Recombinant Human 14-3-3 epsilon Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) PAGE
Formulation In PBS (pH7.4) containing 1mM DTT, 10% glycerol
Molecular Weight (g/mol) 31.3 kDa
Name YWHAE Recombinant Protein
Purification Method >90% by SDS-PAGE
Quantity 100 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias 14-3-3 alpha, 14-3-3 beta, 14-3-3 protein beta/alpha
Gene Symbol YWHAE
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.