Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Activin RIA/ALK-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB637100
This item is not returnable.
View return policy
Description
Activin RIA/ALK-2/Activin Receptor Type 1 Monoclonal specifically detects Activin RIA/ALK-2/Activin Receptor Type 1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Activin RIA/ALK-2/Activin Receptor Type 1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
activin A receptor, type I, ActivinRIA, ACTRI, ACVR1, ACVR1A, ACVRLK2, ALK2, ALK-2, FOP, SKR1, TSRI | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
90 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Immunocytochemistry | |
71412 | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q04771 | |
ACVR1 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Activin RIA/ALK-2 Asp23-Val124 Accession # Q04771 | |
100 μg | |
Primary | |
Detects human Activin RIA/ALK-2 in direct ELISAs and Western blots. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction