Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Activin RIA/ALK-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$200.50 - $2020.00
Specifications
Antigen | Activin RIA/ALK-2/Activin Receptor Type 1 |
---|---|
Clone | 71412 |
Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Description
Activin RIA/ALK-2/Activin Receptor Type 1 Monoclonal specifically detects Activin RIA/ALK-2/Activin Receptor Type 1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Activin RIA/ALK-2/Activin Receptor Type 1 | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
activin A receptor, type I, ActivinRIA, ACTRI, ACVR1, ACVR1A, ACVRLK2, ALK2, ALK-2, FOP, SKR1, TSRI | |
ACVR1 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
71412 | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q04771 | |
90 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Activin RIA/ALK-2 Asp23-Val124 Accession # Q04771 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Activin RIA/ALK-2 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title