Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Activin RIIA/B Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $655.00
Specifications
| Antigen | Activin RIIA/B |
|---|---|
| Clone | 60401 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Activin RIIA/B Monoclonal specifically detects Activin RIIA/B in Human samples. It is validated for Western Blot.Specifications
| Activin RIIA/B | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ActivinRIIA, ActivinRIIB, ACTRII, ACVR2, ACVR2A, ACVR2B, AVR2A | |
| ACVR2A | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 60401 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q13705 | |
| 92 | |
| Mouse myeloma cell line NS0-derived recombinant human Activin RIIB Ser19-Thr134 Accession # Q13705 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Activin RII in direct ELISA and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title