Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ADAM12 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$200.50 - $514.50
Specifications
Antigen | ADAM12 |
---|---|
Clone | 632525 |
Concentration | LYOPH |
Dilution | Immunoprecipitation 25 ug/mL |
Applications | Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB44161SP
![]() |
R&D Systems
MAB44161SP |
25 μg |
Each for $200.50
|
|
|||||
MAB44161
![]() |
R&D Systems
MAB44161 |
100 μg |
Each for $514.50
|
|
|||||
Description
ADAM12 Monoclonal specifically detects ADAM12 in Human samples. It is validated for Immunoprecipitation.Specifications
ADAM12 | |
LYOPH | |
Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
O43184 | |
8038 | |
Chinese hamster ovary cell line CHO-derived recombinant human ADAM12 Arg29-Ser513 Accession # O43184 | |
Primary | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Detects human ADAM12 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human ADAM8, 9, 10, 15, 17, 19, 22, 23, 32, or 33 is observed. |
632525 | |
Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
a disintegrin and metalloproteinase domain 12 (meltrin alpha), ADAM 12, ADAM metallopeptidase domain 12, disintegrin and metalloproteinase domain-containing protein 12, EC 3.4.24, EC 3.4.24.58, MCMP, MCMPMltna, Meltrin alpha, Meltrin-alpha, MLTNA, MLTNEC 3.4.24.- | |
ADAM12 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title