Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ADAMTS3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5465SP
This item is not returnable.
View return policy
Description
ADAMTS3 Polyclonal specifically detects ADAMTS3 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ADAMTS3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
A disintegrin and metalloproteinase with thrombospondin motifs 3, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 3, ADAM metallopeptidase with thrombospondin type 1 motif, 3, ADAM-TS 3, ADAM-TS3, ADAMTS-3, ADAMTS-4, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, KIAA0366PC II-NP, PC II-NP, Procollagen II amino propeptide-processing enzyme, Procollagen II N-proteinase, zinc metalloendopeptidase | |
Sheep | |
Affinity Purified | |
RUO | |
9508 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
O15072 | |
ADAMTS3 | |
Chinese hamster ovary cell line CHO-derived recombinant human ADAMTS3 Ser19-Cys712 Accession # O15072 | |
25 μg | |
Primary | |
Detects human ADAMTS3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 3% cross-reactivity with recombinant human (rh) ADAMTS4 and less than 1% cross-reactivity with rhADAMTS1, rhADAMTS5, rhADAMTS13, and rhADAMTSL1.2 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction