Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ADAMTS3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | ADAMTS3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
ADAMTS3 Polyclonal specifically detects ADAMTS3 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ADAMTS3 | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
A disintegrin and metalloproteinase with thrombospondin motifs 3, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 3, ADAM metallopeptidase with thrombospondin type 1 motif, 3, ADAM-TS 3, ADAM-TS3, ADAMTS-3, ADAMTS-4, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, KIAA0366PC II-NP, PC II-NP, Procollagen II amino propeptide-processing enzyme, Procollagen II N-proteinase, zinc metalloendopeptidase | |
ADAMTS3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
O15072 | |
9508 | |
Chinese hamster ovary cell line CHO-derived recombinant human ADAMTS3 Ser19-Cys712 Accession # O15072 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human ADAMTS3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 3% cross-reactivity with recombinant human (rh) ADAMTS4 and less than 1% cross-reactivity with rhADAMTS1, rhADAMTS5, rhADAMTS13, and rhADAMTSL1.2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title