Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Adenosine A1R Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | Adenosine A1R |
---|---|
Clone | 1051024 |
Dilution | Immunohistochemistry 3-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11135SP
![]() |
R&D Systems
MAB11135SP |
25 μg |
Each for $200.50
|
|
|||||
MAB11135100
![]() |
R&D Systems
MAB11135100 |
100 μg |
Each for $514.50
|
|
|||||
Description
Adenosine A1R Monoclonal antibody specifically detects Adenosine A1R in Human samples. It is validated for ImmunohistochemistrySpecifications
Adenosine A1R | |
Immunohistochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Adenosine A1 R, adenosine A1 receptor, ADORA 1, ADORA1, RDC7, RDC7adenosine receptor A1 | |
Human Adora-1 synthetic peptide corresponding to the 2nd. extra-cellular loop.,, Accession # P30542 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1051024 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P30542 | |
134 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title