Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Afadin/AF-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB78291SP
This item is not returnable.
View return policy
Description
Afadin/AF-6 Monoclonal specifically detects Afadin/AF-6 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Afadin/AF-6 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AF6, AF-6, AF64, AFAD, afadin, FLJ34371, MLLT4, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 4 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4301 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Immunocytochemistry | |
851204 | |
Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P55196 | |
MLLT4 | |
E. coli-derived recombinant human Afadin/AF-6 Asn217-Pro392 Accession # P55196 | |
25 μg | |
Primary | |
Detects human Afadin/AF-6 in direct ELISAs. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction