Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Afadin/AF-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Afadin/AF-6 |
---|---|
Clone | 851204 |
Dilution | Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB78291SP
![]() |
R&D Systems
MAB78291SP |
25 μg |
Each for $200.50
|
|
|||||
MAB78291
![]() |
R&D Systems
MAB78291 |
100 μg |
Each for $502.50
|
|
|||||
Description
Afadin/AF-6 Monoclonal specifically detects Afadin/AF-6 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Afadin/AF-6 | |
Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AF6, AF-6, AF64, AFAD, afadin, FLJ34371, MLLT4, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 4 | |
MLLT4 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
851204 | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P55196 | |
4301 | |
E. coli-derived recombinant human Afadin/AF-6 Asn217-Pro392 Accession # P55196 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Afadin/AF-6 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title