Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ALK-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3701100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3701100 100 μg
MAB3701SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3701100 Supplier R&D Systems Supplier No. MAB3701100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ALK-1 Monoclonal specifically detects ALK-1 in Human samples. It is validated for Blockade of Receptor-ligand Interaction.

Specifications

Antigen ALK-1
Applications Blocking Assay
Classification Monoclonal
Clone 117702
Conjugate Unconjugated
Dilution Blockade of Receptor-ligand Interaction 0.15-0.75 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P37023
Gene Alias activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRL1, ACVRLK1ORW2, ALK1, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ALK-1, Asp22-Gln118, Accession # P37023
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 94
Test Specificity Detects human ALK-1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.