Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ALK-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | ALK-1 |
---|---|
Clone | 117702 |
Dilution | Blockade of Receptor-ligand Interaction 0.15-0.75 μg/mL |
Applications | Blocking Assay |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3701SP
![]() |
R&D Systems
MAB3701SP |
25 μg |
Each for $200.50
|
|
|||||
MAB3701100
![]() |
R&D Systems
MAB3701100 |
100 μg |
Each for $479.50
|
|
|||||
Description
ALK-1 Monoclonal specifically detects ALK-1 in Human samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
ALK-1 | |
Blockade of Receptor-ligand Interaction 0.15-0.75 μg/mL | |
Monoclonal | |
Lyophilized | |
Human | |
P37023 | |
94 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
117702 | |
Blocking Assay | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRL1, ACVRLK1ORW2, ALK1, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I | |
Mouse myeloma cell line NS0-derived recombinant human ALK-1, Asp22-Gln118, Accession # P37023 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ALK-1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title