Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human alpha-Fetoprotein/AFP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB13691
This item is not returnable.
View return policy
Description
alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for Western Blot.Specifications
alpha-Fetoprotein/AFP | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
AFP, Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, DSCAM2, FETA, HP, HPAFP | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
174 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot | |
214107 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AFP | |
Human umbilical cord serum-derived alpha-Fetoprotein/AFP | |
500 μg | |
Primary | |
Detects purified human alpha-Fetoprotein/AFP in Western blots. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction