Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human alpha-Fetoprotein/AFP Antibody, R&D Systems™

Mouse Monoclonal Antibody
$208.00 - $679.50
Specifications
Antigen | alpha-Fetoprotein/AFP |
---|---|
Clone | 214107 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB13691SP
![]() |
R&D Systems
MAB13691SP |
25 μg |
Each for $208.00
|
|
|||||
MAB13691
![]() |
R&D Systems
MAB13691 |
500 μg |
Each for $679.50
|
|
|||||
Description
alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for Western Blot.Specifications
alpha-Fetoprotein/AFP | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
AFP, Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, DSCAM2, FETA, HP, HPAFP | |
AFP | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
214107 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
174 | |
Human umbilical cord serum-derived alpha-Fetoprotein/AFP | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects purified human alpha-Fetoprotein/AFP in Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title