Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human alpha-Synuclein Antibody, R&D Systems™

Mouse Monoclonal Antibody
$177.10 - $454.00
Specifications
Antigen | alpha-Synuclein |
---|---|
Clone | 1060506 |
Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL |
Applications | Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11376100
![]() |
R&D Systems
MAB11376100 |
100 μg |
Each for $454.00
|
|
|||||
MAB11376SP
![]() |
R&D Systems
MAB11376SP |
25 μg |
Each for $177.10
|
|
|||||
Description
alpha-Synuclein Monoclonal antibody specifically detects alpha-Synuclein in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
alpha-Synuclein | |
Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), Synuclein-alpha, truncated alpha synuclein | |
E. coli -derived human alpha-Synuclein, Met1-Ala140, Accession # P37840 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1060506 | |
Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P37840 | |
6622 | |
IgG2b | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title