Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human AMIGO2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2080SP
This item is not returnable.
View return policy
Description
AMIGO2 Polyclonal specifically detects AMIGO2 in Human samples. It is validated for Western Blot.Specifications
AMIGO2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
adhesion molecule with Ig-like domain 2, ALI1amphoterin induced gene 2, alivin 1, Alivin-1, AMIGO-2, DEGAamphoterin-induced protein 2, differentially expressed in gastric adenocarcinoma, Differentially expressed in gastric adenocarcinomas, transmembrane protein AMIGO2 | |
Goat | |
Affinity Purified | |
RUO | |
347902 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Q86SJ2 | |
AMIGO2 | |
Mouse myeloma cell line NS0-derived recombinant human AMIGO2 Gly39-Ala392 Accession # Q86SJ2 | |
25 μg | |
Primary | |
Detects human AMIGO2 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human AMIGO and rhAMIGO3 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction