Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human AMIGO2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $538.00
Specifications
Antigen | AMIGO2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
AMIGO2 Polyclonal specifically detects AMIGO2 in Human samples. It is validated for Western Blot.Specifications
AMIGO2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
adhesion molecule with Ig-like domain 2, ALI1amphoterin induced gene 2, alivin 1, Alivin-1, AMIGO-2, DEGAamphoterin-induced protein 2, differentially expressed in gastric adenocarcinoma, Differentially expressed in gastric adenocarcinomas, transmembrane protein AMIGO2 | |
AMIGO2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q86SJ2 | |
347902 | |
Mouse myeloma cell line NS0-derived recombinant human AMIGO2 Gly39-Ala392 Accession # Q86SJ2 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human AMIGO2 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human AMIGO and rhAMIGO3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title