Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human AMPKβ2 Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
$182.85 - $449.22
Specifications
Antigen | AMPK beta 2/PRKAB2 |
---|---|
Clone | 1092615 |
Dilution | Western Blot 2 μg/mL, Simple Western 20 μg/mL |
Applications | Western Blot, Simple Western |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11639100
|
R&D Systems
MAB11639100 |
100 μg |
Each for $449.22
|
|
|||||
MAB11639SP
|
R&D Systems
MAB11639SP |
25 μg |
Each for $182.85
|
|
|||||
Description
AMPK beta 2/PRKAB2 Monoclonal antibody specifically detects AMPK beta 2/PRKAB2 in Human samples. It is validated for Western Blot,Simple WesternSpecifications
AMPK beta 2/PRKAB2 | |
Western Blot 2 μg/mL, Simple Western 20 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
5'-AMP-activated protein kinase, beta-2 subunit, AMPK beta 2,5'-AMP-activated protein kinase subunit beta-2, AMPK beta-2 chain, AMPK subunit beta-2, MGC61468, PRKAB2, protein kinase, AMP-activated, beta 2 non-catalytic subunit | |
E. coli - derived recombinant human AMPKbeta2, Met1-Ile272, Accession # O43741 | |
Primary | |
Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration. |
1092615 | |
Western Blot, Simple Western | |
Unconjugated | |
Mouse | |
Human | |
O43741 | |
5565 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°CC as supplied. 1 month, 2 to 8°CC under sterile conditions after reconstitution. 6 months, -20 to -70°CC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title