Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Angiopoietin-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $643.50
Specifications
| Antigen | Angiopoietin-4 |
|---|---|
| Clone | 156210 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
Description
Angiopoietin-4 Monoclonal specifically detects Angiopoietin-4 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| Angiopoietin-4 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGP4, ANG3, ANG-3, ANG-4, ANG4angiopoietin-3, angiopoietin 4, Angiopoietin-3, angiopoietin-4, ANGPT4, dJ824F16.2 (angiopoietin 4), MGC138181, MGC138183 | |
| ANGPT4 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 156210 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9Y264 | |
| 51378 | |
| Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-4 Met1-Ile503 Accession # Q9Y264 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects human Angiopoietin-4 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Angiopoietin-1, -2, rhAngiopoietin-like 3, rhAngiopoietin-like 4, rhTie-2, or recombinant mouse Angiopoietin-3 was observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title