Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Annexin V Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3991SP
This item is not returnable.
View return policy
Description
Annexin V Monoclonal specifically detects Annexin V in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.Specifications
Annexin V | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, ANXA5, ANXV, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha | |
E. coli-derived recombinant human Annexin V Met1-Asp320 Accession # P08758 | |
25 μg | |
Primary | |
Detects human Annexin V in direct ELISAs. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
38701 | |
Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunohistochemistry 5-25 ug/mL | |
P08758 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
308 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction