Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Annexin V Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Annexin V |
---|---|
Clone | 38701 |
Dilution | Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunohistochemistry 5-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
MAB3991SP
![]() |
R&D Systems
MAB3991SP |
Unconjugated | 25 μg |
Each for $200.50
|
|
||||
MAB3991100
![]() |
R&D Systems
MAB3991100 |
Unconjugated | 100 μg |
Each for $549.50
|
|
||||
Description
Annexin V Monoclonal specifically detects Annexin V in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.Specifications
Annexin V | |
Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, ANXA5, ANXV, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha | |
E. coli-derived recombinant human Annexin V Met1-Asp320 Accession # P08758 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Annexin V in direct ELISAs. |
38701 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P08758 | |
308 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title