Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Apolipoprotein A-I/ApoA1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF3664SP
This item is not returnable.
View return policy
Description
Apolipoprotein A-I/ApoA1 Polyclonal specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for Western Blot, Simple Western.Specifications
Apolipoprotein A-I/ApoA1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Alp-1, APOA1, Apo-AI, ApoA-I, Apolipoprotein A1, apolipoprotein A-I, Apolipoprotein AI, Brp-14, Ltw-1, Lvtw-1, MGC117399, Sep-1, Sep-2 | |
Goat | |
Affinity Purified | |
RUO | |
335 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL | |
P02647 | |
APOA1 | |
Human Plasma ApoA1 and E. coli-derived recombinant human ApoA1 Asp25-Gln267 Accession # P02647 | |
25 μg | |
Primary | |
Detects human ApoA1 in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) ApoA2, rhApoB100, and rhApoC2 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction