Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Apolipoprotein A-I/ApoA1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$214.00 - $585.00
Specifications
Antigen | Apolipoprotein A-I/ApoA1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.5 ug/mL, Simple Western 5 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Apolipoprotein A-I/ApoA1 Polyclonal specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for Western Blot, Simple Western.Specifications
Apolipoprotein A-I/ApoA1 | |
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Alp-1, APOA1, Apo-AI, ApoA-I, Apolipoprotein A1, apolipoprotein A-I, Apolipoprotein AI, Brp-14, Ltw-1, Lvtw-1, MGC117399, Sep-1, Sep-2 | |
APOA1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P02647 | |
335 | |
Human Plasma ApoA1 and E. coli-derived recombinant human ApoA1 Asp25-Gln267 Accession # P02647 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human ApoA1 in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) ApoA2, rhApoB100, and rhApoC2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title