Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Apolipoprotein A-II/ApoA2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Apolipoprotein A-II/ApoA2 |
---|---|
Clone | 395906 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Apolipoprotein A-II/ApoA2 Monoclonal specifically detects Apolipoprotein A-II/ApoA2 in Human samples. It is validated for Western Blot.Specifications
Apolipoprotein A-II/ApoA2 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Human | |
P02652 | |
336 | |
E. coli-derived recombinant human ApoA2 Gln24-Gln100 Accession # P02652 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ApoA2 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) ApoA1 or rhApoE. |
395906 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Alp-2, APOA2, apoAII, Apo-AII, ApoA-II, Apolipoprotein A2, apolipoprotein A-II, Apolipoprotein AII, Hdl-1 | |
APOA2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title