Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human APP695+1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB890
This item is not returnable.
View return policy
Description
APP 695+1 Monoclonal specifically detects APP 695+1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
APP 695+1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
APP | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human APP695 +1 frameshift mutant Leu18-Arg336 (predicted) Accession # NP_958817 | |
500 μg | |
Primary | |
Detects human APP695 +1 in Western blots. It recognizes a common epitope present on APP695 +1 and APP770 found within the segments L18-R288 or P365-R411 of APP770 (Accession # P05067). | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
128715 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
NP_958817 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
351 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction