Learn More
Human ATAD2 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | ATAD2 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 952947 |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | Q6PL18 |
| Gene Alias | AAA nuclear coregulator cancer-associated protein, ANCCA, ANCCAMGC131938, ATPase family AAA domain-containing protein 2, ATPase family, AAA domain containing 2, CT137, DKFZp667N1320, EC 3.6.1.3, MGC142216, MGC29843, MGC5254, PRO2000 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.