Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Bcl-2 Minus C-Terminus Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8271
This item is not returnable.
View return policy
Description
Bcl-2 Monoclonal specifically detects Bcl-2 in Human samples. It is validated for Neutralization.Specifications
Bcl-2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BCL2 | |
E. coli-derived recombinant human Bcl-2 minus the carboxyl-terminal mitochondrial targeting sequence | |
100 μg | |
Primary | |
Detects human Bcl-2 Minus C-Terminus. Neutralizes recombinant human (rh) Bcl-2 bioactivity but not rhBcl-w or rhBcl-x bioactivity. | |
Human | |
Purified |
Neutralization | |
118712 | |
Neutralization | |
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl2, Bcl-2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
596 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction