Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Bcl-2 Minus C-Terminus Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $526.50
Specifications
Antigen | Bcl-2 |
---|---|
Clone | 118712 |
Dilution | Neutralization |
Applications | Neutralization |
Classification | Monoclonal |
Description
Bcl-2 Monoclonal specifically detects Bcl-2 in Human samples. It is validated for Neutralization.Specifications
Bcl-2 | |
Neutralization | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
596 | |
E. coli-derived recombinant human Bcl-2 minus the carboxyl-terminal mitochondrial targeting sequence | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Bcl-2 Minus C-Terminus. Neutralizes recombinant human (rh) Bcl-2 bioactivity but not rhBcl-w or rhBcl-x bioactivity. |
118712 | |
Neutralization | |
Unconjugated | |
Mouse | |
Human | |
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl2, Bcl-2 | |
BCL2 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title