Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human beta-1 Adrenergic R/ADRB1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | beta-1 Adrenergic R/ADRB1 |
---|---|
Clone | 2203D |
Dilution | Western Blot 1 μ/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10119SP
![]() |
R&D Systems
MAB10119SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10119100
![]() |
R&D Systems
MAB10119100 |
100 μg |
Each for $491.00
|
|
|||||
Description
beta-1 Adrenergic R/ADRB1 Monoclonal specifically detects beta-1 Adrenergic R/ADRB1 in Human samples. It is validated for Western Blot.Specifications
beta-1 Adrenergic R/ADRB1 | |
Western Blot 1 μ/mL | |
Monoclonal | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ADRB1, ADRB1R, ADRB1RRHR, adrenergic, beta-1-, receptor, B1AR, beta-1 adrenergic receptor, beta1 AdrenergicR, beta-1 AdrenergicR, Beta-1 adrenoceptor, Beta-1 adrenoreceptor, BETA1AR | |
Synthetic peptide containing human beta-1 Adrenergic R/ADRB1, Accession No. P08588 | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
2203D | |
Western Blot | |
Unconjugated | |
Human | |
P08588 | |
153 | |
IgG | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human beta-1 Adrenergic R/ADRB1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title