Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human beta-3 Adrenergic R/ADRB3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $549.50
Specifications
Antigen | beta-3 Adrenergic R/ADRB3 |
---|---|
Clone | 493324 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL |
Applications | Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9419SP
![]() |
R&D Systems
MAB9419SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9419100
![]() |
R&D Systems
MAB9419100 |
100 μg |
Each for $549.50
|
|
|||||
Description
beta-3 Adrenergic R/ADRB3 Monoclonal specifically detects beta-3 Adrenergic R/ADRB3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
beta-3 Adrenergic R/ADRB3 | |
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ADRB3, ADRB3R, adrenergic, beta-3-, receptor, B3AR, beta-3 adrenergic receptor, beta3 AdrenergicR, beta-3 AdrenergicR, Beta-3 adrenoceptor, Beta-3 adrenoreceptor, BETA3AR, FLJ99960 | |
Human embryonic kidney cell line HEK293-derived transfected with human beta-3 Adrenergic R/ADRB3 Met1-Ser408 Accession # P13945 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Stains human beta-3 Adrenergic R/ADRB3 transfectants but not irrelevent transfectants in flow cytometry. |
493324 | |
Flow Cytometry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P13945 | |
155 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title