Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMP-5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB715
This item is not returnable.
View return policy
Description
BMP-5 Monoclonal specifically detects BMP-5 in Human samples. It is validated for Western Blot.Specifications
BMP-5 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP5, BMP-5, bone morphogenetic protein 5, MGC34244 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
653 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
78205 | |
Western Blot 1 ug/mL | |
P22003 | |
BMP5 | |
Mouse myeloma cell line NS0-derived recombinant human BMP-5 Accession # P22003 | |
500 μg | |
Primary | |
Detects human BMP-5 in direct ELISAs and Western blots. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction