Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMP-5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | BMP-5 |
---|---|
Clone | 78205 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
BMP-5 Monoclonal specifically detects BMP-5 in Human samples. It is validated for Western Blot.Specifications
BMP-5 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP5, BMP-5, bone morphogenetic protein 5, MGC34244 | |
BMP5 | |
IgG2a | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
78205 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P22003 | |
653 | |
Mouse myeloma cell line NS0-derived recombinant human BMP-5 Accession # P22003 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human BMP-5 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title