Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BPI Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7468SP
This item is not returnable.
View return policy
Description
BPI Monoclonal specifically detects BPI in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
BPI | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
bactericidal permeability-increasing protein, bactericidal/permeability-increasing protein, BPIFD1, CAP 57, rBPI | |
HEK293 human embryonic kidney cell line transfected with human BPI Val32-Lys487 Accession # P17213 | |
25 μg | |
Primary | |
Detects human BPI in direct ELISAs. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
971513 | |
Western Blot 1 ug/mL, Immunohistochemistry 1-25 ug/mL | |
P17213 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
671 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction