Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BPI Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $526.50
Specifications
Antigen | BPI |
---|---|
Clone | 971513 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 1-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
MAB7468SP
![]() |
R&D Systems
MAB7468SP |
Unconjugated | 25 μg |
Each for $200.50
|
|
||||
MAB7468100
![]() |
R&D Systems
MAB7468100 |
Unconjugated | 100 μg |
Each for $526.50
|
|
||||
Description
BPI Monoclonal specifically detects BPI in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
BPI | |
Western Blot 1 ug/mL, Immunohistochemistry 1-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
bactericidal permeability-increasing protein, bactericidal/permeability-increasing protein, BPIFD1, CAP 57, rBPI | |
HEK293 human embryonic kidney cell line transfected with human BPI Val32-Lys487 Accession # P17213 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human BPI in direct ELISAs. |
971513 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P17213 | |
671 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title