Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BTN1A1/Butyrophilin Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

$214.00 - $538.00

Specifications

Antigen BTN1A1/Butyrophilin
Clone 2151B
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 0.3-25 ug/mL
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB84671SP
GREENER_CHOICE
View Documents
R&D Systems
MAB84671SP
25 μg
Each for $214.00
Only null left
Add to Cart
 
MAB84671100
GREENER_CHOICE
View Documents
R&D Systems
MAB84671100
100 μg
Each for $538.00
Only null left
Add to Cart
 
Description

Description

BTN1A1/Butyrophilin Monoclonal specifically detects BTN1A1/Butyrophilin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications

Specifications

BTN1A1/Butyrophilin
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 0.3-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), BTN, BTN1A1, butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1
Mouse myeloma cell line NS0-derived recombinant human BTN1A1/Butyrophilin Ala27-Arg242 Accession # Q13410
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human BTN1A1/Butyrophilin in direct ELISAs.
2151B
Flow Cytometry, Immunocytochemistry
Unconjugated
Rabbit
Human
Q13410
696
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only (RUO)

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.