Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cadherin-12 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2240
This item is not returnable.
View return policy
Description
Cadherin-12 Polyclonal specifically detects Cadherin-12 in Human samples. It is validated for Western Blot.Specifications
Cadherin-12 | |
Polyclonal | |
Unconjugated | |
P55289 | |
CDH12 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Cadherin-12 Gly55-Pro609 Accession # P55289 | |
100 μg | |
Primary | |
Detects human Cadherin-12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) Cadherin-8, rhCadherin-11, rhN-Cadherin, and rhVE-Cadherin is observed and less than 1% cross-reactivity with rhE-Cadherin, rhP-Cadherin, rhCadherin-17, and rhCadherin-4 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
BR-cadherin, cadherin 12, type 2 (N-cadherin 2), cadherin-12, Cadherin12, CDH12, CDHB, FLJ34857, N-Cadherin 2, Neural type cadherin 2, neural type, 2, neuronal cadherin 2 | |
Goat | |
Affinity Purified | |
RUO | |
1010 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction