Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cadherin-12 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | Cadherin-12 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Cadherin-12 Polyclonal specifically detects Cadherin-12 in Human samples. It is validated for Western Blot.Specifications
Cadherin-12 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
BR-cadherin, cadherin 12, type 2 (N-cadherin 2), cadherin-12, Cadherin12, CDH12, CDHB, FLJ34857, N-Cadherin 2, Neural type cadherin 2, neural type, 2, neuronal cadherin 2 | |
CDH12 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P55289 | |
1010 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Cadherin-12 Gly55-Pro609 Accession # P55289 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Cadherin-12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) Cadherin-8, rhCadherin-11, rhN-Cadherin, and rhVE-Cadherin is observed and less than 1% cross-reactivity with rhE-Cadherin, rhP-Cadherin, rhCadherin-17, and rhCadherin-4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title