Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cadherin-17 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$200.50 - $2020.00
Specifications
Antigen | Cadherin-17 |
---|---|
Clone | 141713 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 0.5-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1032SP
![]() |
R&D Systems
MAB1032SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1032100
![]() |
R&D Systems
MAB1032100 |
100 μg |
Each for $538.00
|
|
|||||
MAB1032500
![]() |
R&D Systems
MAB1032500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
Cadherin-17 Monoclonal specifically detects Cadherin-17 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cadherin-17 | |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cadherin, cadherin 17, LI cadherin (liver-intestine), cadherin-16, cadherin-17, Cadherin17, CDH16, CDH17, FLJ26931, HPT1, HPT-1, HPT-1 cadherin, human intestinal peptide-associated transporter HPT-1, human peptide transporter 1, Intestinal peptide-associated transporter HPT-1, LI cadherin, LI-cadherin, Liver-intestine cadherin, MGC138218, MGC142024 | |
CDH17 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
141713 | |
Western Blot 1 ug/mL, Immunohistochemistry 0.5-25 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
CAA58231 | |
1015 | |
Mouse myeloma cell line NS0-derived recombinant human Cadherin-17 Gln23-Met787 Accession # CAA58231 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Cadherin-17 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) Cadherin-8, rhE-Cadherin, rhP-Cadherin, or rhVE-Cadherin. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title