Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CaM Kinase II alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5584SP
This item is not returnable.
View return policy
Description
CaM Kinase II alpha Monoclonal specifically detects CaM Kinase II alpha in Human samples. It is validated for Immunohistochemistry.Specifications
CaM Kinase II alpha | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
Q9UQM7 | |
CAMK2A | |
E. coli-derived recombinant human CaM Kinase II alpha Ala144-Ala262 Accession # Q9UQM7 | |
25 μg | |
Primary | |
Detects human CaM Kinase II alpha in direct ELISAs. | |
Human | |
Purified |
Immunohistochemistry | |
525501 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, CaM kinase II alpha subunit, CaM kinase II subunit alpha, CAMK2A, CAMKAcalcium/calmodulin-dependent protein kinase type II subunit alpha, CaMK-II alpha subunit, CaMK-II subunit alpha, CaMKIINalpha, CaM-kinase II alpha chain, EC 2.7.11, EC 2.7.11.17, KIAA0968calcium/calmodulin-dependent protein kinase type II alpha chain | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
815 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction