Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CaM Kinase II alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | CaM Kinase II alpha |
---|---|
Clone | 525501 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Description
CaM Kinase II alpha Monoclonal specifically detects CaM Kinase II alpha in Human samples. It is validated for Immunohistochemistry.Specifications
CaM Kinase II alpha | |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q9UQM7 | |
815 | |
E. coli-derived recombinant human CaM Kinase II alpha Ala144-Ala262 Accession # Q9UQM7 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CaM Kinase II alpha in direct ELISAs. |
525501 | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, CaM kinase II alpha subunit, CaM kinase II subunit alpha, CAMK2A, CAMKAcalcium/calmodulin-dependent protein kinase type II subunit alpha, CaMK-II alpha subunit, CaMK-II subunit alpha, CaMKIINalpha, CaM-kinase II alpha chain, EC 2.7.11, EC 2.7.11.17, KIAA0968calcium/calmodulin-dependent protein kinase type II alpha chain | |
CAMK2A | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title