Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carboxypeptidase E/CPE Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems MAB3587
This item is not returnable.
View return policy
Description
Carboxypeptidase E/CPE Monoclonal specifically detects Carboxypeptidase E/CPE in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carboxypeptidase E/CPE | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
P16870 | |
CPE | |
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase E/CPE Arg42-Ser453 Accession # P16870 | |
100 μg | |
Primary | |
Detects human Carboxypeptidase E/CPE in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CPA1, rhCPA2, rhCPB1, or recombinant mouse CPA4 is observed. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
420709 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
carboxypeptidase E, Carboxypeptidase H, CPE, CPH, CPH-1, EC 3.4.17, EC 3.4.17.10, Enkephalin convertase, Prohormone-processing carboxypeptidase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1363 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction