Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carboxypeptidase E/CPE Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
$200.50 - $502.50
Specifications
Antigen | Carboxypeptidase E/CPE |
---|---|
Clone | 420709 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Carboxypeptidase E/CPE Monoclonal specifically detects Carboxypeptidase E/CPE in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carboxypeptidase E/CPE | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
P16870 | |
1363 | |
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase E/CPE Arg42-Ser453 Accession # P16870 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Carboxypeptidase E/CPE in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CPA1, rhCPA2, rhCPB1, or recombinant mouse CPA4 is observed. |
420709 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
carboxypeptidase E, Carboxypeptidase H, CPE, CPH, CPH-1, EC 3.4.17, EC 3.4.17.10, Enkephalin convertase, Prohormone-processing carboxypeptidase | |
CPE | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title