Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carboxypeptidase X1/CPXM1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Carboxypeptidase X1/CPXM1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
Carboxypeptidase X1/CPXM1 Polyclonal specifically detects Carboxypeptidase X1/CPXM1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carboxypeptidase X1/CPXM1 | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Human | |
carboxypeptidase X (M14 family), carboxypeptidase X (M14 family), member 1, CPX1, CPX-1, CPX1EC 3.4.17.-, CPXM, CPXM1, EC 3.4.17, EC 3.4.17.10, EC 3.4.17.3, Metallocarboxypeptidase CPX-1, probable carboxypeptidase X1 | |
CPXM1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
Q96SM3 | |
56265 | |
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase X1/CPXM1 Ala21-Asp734 Accession # Q96SM3 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Carboxypeptidase X1/CPXM1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse CPXM is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title