Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carnosine Dipeptidase 1/CNDP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | Carnosine Dipeptidase 1/CNDP1 |
---|---|
Clone | 352213 |
Concentration | LYOPH |
Dilution | Immunoprecipitation 25 ug/mL, Neutralization 0.6 ug/mL |
Applications | Immunoprecipitation, Neutralization |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB24891SP
![]() |
R&D Systems
MAB24891SP |
25 μg |
Each for $200.50
|
|
|||||
MAB24891
![]() |
R&D Systems
MAB24891 |
100 μg |
Each for $468.00
|
|
|||||
Description
Carnosine Dipeptidase 1/CNDP1 Monoclonal specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for Immunoprecipitation, Neutralization.Specifications
Carnosine Dipeptidase 1/CNDP1 | |
LYOPH | |
Immunoprecipitation, Neutralization | |
Unconjugated | |
Mouse | |
Human | |
Q96KN2 | |
84735 | |
Mouse myeloma cell line NS0-derived recombinant human Carnosine Dipeptidase 1/CNDP1 Pro28-His507 Accession # Q96KN2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Carnosine Dipeptidase 1/CNDP1 in direct ELISAs. |
352213 | |
Immunoprecipitation 25 ug/mL, Neutralization 0.6 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CNDP1, CPGL2, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase | |
CNDP1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title