Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Caspase-3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB7071SP
This item is not returnable.
View return policy
Description
Caspase-3 Monoclonal specifically detects Caspase-3 in Human samples. It is validated for Immunocytochemistry, Dual RNAscope ISH-IHC.Specifications
Caspase-3 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
836 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Immunocytochemistry, In Situ Hybridization (ISH), Immunohistochemistry | |
1112I | |
Immunocytochemistry 1-25 ug/mL, Dual RNAscope ISH-IHC 5-25 ug/mL | |
P42574 | |
CASP3 | |
KLH-conjugated human Caspase-3 synthetic peptide, CRGTELDCGIETD, Accession # P42574 | |
25 μg | |
Primary | |
Detects the active form and p17 subunit of Caspase-3 in direct ELISAs, with less than 5% cross-reactivity with the precursor form observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction