Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Caspase-3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$201.00 - $410.00
Specifications
Antigen | Caspase-3 |
---|---|
Clone | 1112I |
Dilution | Immunocytochemistry 1-25 ug/mL, Dual RNAscope ISH-IHC 5-25 ug/mL |
Applications | Immunocytochemistry, In Situ Hybridization (ISH), Immunohistochemistry |
Classification | Monoclonal |
Description
Caspase-3 Monoclonal specifically detects Caspase-3 in Human samples. It is validated for Immunocytochemistry, Dual RNAscope ISH-IHC.Specifications
Caspase-3 | |
Immunocytochemistry 1-25 ug/mL, Dual RNAscope ISH-IHC 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
CASP3 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
1112I | |
Immunocytochemistry, In Situ Hybridization (ISH), Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P42574 | |
836 | |
KLH-conjugated human Caspase-3 synthetic peptide, CRGTELDCGIETD, Accession # P42574 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects the active form and p17 subunit of Caspase-3 in direct ELISAs, with less than 5% cross-reactivity with the precursor form observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title