Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human Caspase-9 ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Click to view available options
Quantity:
96 Tests
Description
The Human Caspase-9 ELISA Kit (Colorimetric) measures human Caspase-9 in cell culture supernatant, cell lysate, serum.
Specifications
Specifications
Assay Range | 1.6 - 100ng/mL |
Assay Sensitivity | 0.4ng/mL |
Product Type | ELISA Kit |
Sample Type | Cell Culture Supernatant, Cell Lysates, Serum |
Specificity | The interference of circulating factors of the immune system was evaluated by spiking these proteins at physiologically relevant concentrations into a human Caspase 9 positive sample. No crossreactivity was detected. |
Target Species | Human |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 842 |
Gene Symbol | CASP9 |
Kit Contents | Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human Caspase-9, Anti-human Caspase-9 polyclonal Detection Antibody (rabbit), Anti-rabbit-IgG-HRP, Human Caspase-9 Standard lyophilized, 200ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Lysis Buffer 10x, Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction