Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cathepsin E Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1294
This item is not returnable.
View return policy
Description
Cathepsin E Monoclonal specifically detects Cathepsin E in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cathepsin E | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
NP_001901 | |
CTSE | |
Mouse myeloma cell line NS0-derived recombinant human Cathepsin E Gln18-Pro396 Accession # NP_001901 | |
500 μg | |
Primary | |
Detects human Cathepsin E in direct ELISAs and Western blots. It recognizes both the pro and mature forms of recombinant human (rh) Cathepsin E. In Western blots, 100% cross-reactivity with recombinant mouse (rm) Cathepsin E and recombinant human (rh) BACE is observed under reducing conditions. Under non-reducing conditions, less than 1% cross-reactivity with rmCathepsin E and rhBACE-1 is observed. Less than 1% cross-reactivity with rhCathepsin D is observed under both reducing and non-reducing conditions. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
212211 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CATE, cathepsin E, CTSE, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1510 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction