Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCK Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB11040SP
This item is not returnable.
View return policy
Description
Cholecystokinin Monoclonal antibody specifically detects Cholecystokinin in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
Cholecystokinin | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
cholecystokinin, MGC117187 | |
Human CCK synthetic peptide, Accession # P06307 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG |
Immunohistochemistry, Immunocytochemistry | |
2753A | |
Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
P06307 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
885 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction